Anti-PHGDH Rabbit Polyclonal Antibody

Leverancier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

61-613
PRSI61-613EA 620 EUR
PRSI61-613
Anti-PHGDH Rabbit Polyclonal Antibody
Antilichamen
For IHC-P starting dilution is: 1:50~100. For FACS starting dilution is: 1:10~50. For WB starting dilution is: 1:1000.

Type: Primary
Antigen: PHGDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 249-277
Host: Rabbit
Isotype: Ig
Reactivity: Human
New Product
Order Now

Specificaties

antigeen symbool PHGDH
Environmentally Preferable
Type antilichaam D-3-phosphoglycerate dehydrogenase,Primair
Type antilichaam D-3-phosphoglycerate dehydrogenase,Primair
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Gen-ID 26227
Isotype
Western blot Yes
ImmunoChemie Yes
Flowcytometrie Yes
Modification None
Epitoop 249-277 aa
Immunogeen This PHGDH antibody is generated from rabbits immunised with a KLH conjugated synthetic peptide between 249-277 amino acids from the central region of human PHGDH.
Formulier Liquid
Moleculair gewicht 57 kDa
Purification This antibody is purified through a protein A column, followed by peptide affinity purification.
Concentratie 0,5 mg/ml
Opslagtemperatuur Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR