Anti-TIMP1 Rabbit Polyclonal Antibody

Leverancier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

62-416
PRSI62-416EA 620 EUR
PRSI62-416
Anti-TIMP1 Rabbit Polyclonal Antibody
Antilichamen
For WB starting dilution is: 1:1000 For IHC-P starting dilution is: 1:10~50 For FACS starting dilution is: 1:10~50 For IF starting dilution is: 1:10~50.

Type: Primary
Antigen: TIMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 41-70
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR