Anti-KLF5 Rabbit Polyclonal Antibody

Leverancier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

62-840
PRSI62-840EA 620 EUR
PRSI62-840
Anti-KLF5 Rabbit Polyclonal Antibody
Antilichamen
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: KLF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 127-156
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR