Human BMP15 peptide
Leverancier: ProSci Inc.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6909P
PRSI6909PEA 122 EUR
PRSI6909P
Human BMP15 peptide
Eiwitten en peptiden
BMP15 peptide is used for blocking the activity of BMP15 antibody.