Human recombinant Fructose-1,6-Bisphosphatase 1 (from E. coli)
Leverancier: ProSci Inc.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
91-122
PRSI91-122EA 567 EUR
PRSI91-122
Human recombinant Fructose-1,6-Bisphosphatase 1 (from E. coli)
Enzymen
Fructose-1,6-Bisphosphatase 1 (FBPase 1) is a member of the FBPase class 1 family. FBPase 1 is a gluconeogenesis regulatory protein, which catalyses the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate. FBPase 1 can assume an active R-state, or an inactive T-state. FBPase 1 deficiency is inherited as an autosomal recessive disorder mainly in the liver and causes life-threatening episodes of hypoglycemia and metabolic acidosis in newborn infants or young children. FBPase 1 coupled with phosphofructokinase (PFK) is involved in the metabolism of pancreatic islet cells.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.