Human recombinant IGF Binding 4 (from cells)
Leverancier: ProSci Inc.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
91-320
PRSI91-320EA 734 EUR
PRSI91-320
Human recombinant IGF Binding 4 (from cells)
Eiwitten en peptiden
IGFBP-4, whose full name is insulin-like growth factor-binding protein 4, is induced by forskolin and N6, O2’dibutyryl sdenosine 3’, or 5’-cyclic monophosphate. It contains IGFBP N-terminal domain and thyroglobulin type-1 domain, and can bind IGF2. The IGF-binding proteins can prolong the half-life of the IGFs and have been shown to either inhibit or stimulate the growth promoting effects of the IGFs on cell culture.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag