Mouse recombinant NOV

Leverancier: ProSci Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

92-639
PRSI92-639EA 461 EUR
PRSI92-639
Mouse recombinant NOV
Eiwitten en peptiden

NOV, also called CCN3, is a secreted protein of CCN family members. CCN family members are highly conserved cysteine rich proteins sharing a common modular structure having 4 conserved domains, insulin-like growth factor-binding protein (IGFBP) domain, von Willebrand type C (VWC) domain, thrombospondin-1 (TSP-1) domain, and C-terminal (CT) domain (absent in CCN5). By specific interactions with these domains, CCN proteins modulate multiple signalling pathways including BMPs, Wnt, TGFs, Notch and integrins to regulate cell proliferation, differentiation, adhesion, migration, angiogenesis, and survival. CCN3 is firstly characterised as a promoter of progenitor activity of human hematopoietic stem cells, as knockdown of CCN3 can abrogate the function of primitive progenitors. Recent studies showed that CCN3 is also actively involved in the process of wound healing. CCN3 is highly expressed in granulation tissues of cutaneous wounds and capable of inducing synthetic responses of fibroblasts.


Fusion-Tag: C-6 His tag


This recombinant protein can be used for biological assays. For research use only.

Order Now

Specificaties

Protein/peptide name NOV
Protein synonyms CCN family member 3|NovH|IGFBP-9|Nov|Protein NOV homolog|Nephroblastoma-overexpressed gene protein homolog|CCN3
Environmentally Preferable
Protein/peptide type Recombinant
Species Mouse
Bron
Sequentie Ser26-Ile354
Zuiverheid > 95% as determined by reducing SDS-PAGE
Formulering Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Moleculair gewicht 39.1 kD
Storage conditions Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
Tag sequence C-6 His tag

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR