Occupational shoes, slip-on, FW58
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FW58WHR48 FW58WHR34 FW58WHR37 FW58WHR42 FW58WHR41 FW58WHR35 FW58WHR44 FW58WHR36 FW58WHR40 FW58WHR38 FW58WHR39 FW58WHR43 FW58WHR47 FW58WHR45 FW58WHR46
PRTWFW58WHR48EA 45 EUR
PRTWFW58WHR48 PRTWFW58WHR34 PRTWFW58WHR37 PRTWFW58WHR42 PRTWFW58WHR41 PRTWFW58WHR35 PRTWFW58WHR44 PRTWFW58WHR36 PRTWFW58WHR40 PRTWFW58WHR38 PRTWFW58WHR39 PRTWFW58WHR43 PRTWFW58WHR47 PRTWFW58WHR45 PRTWFW58WHR46
Occupational shoes, slip-on, FW58
Schoenen en laarzen
Washable upper with a self-cleaning outsole makes this slip on shoe ideal when a protective toecap is not required. Lightweight and comfortable construction.
  • Anti-static footwear
  • Energy absorbing seat region
  • Water resistant upper to prevent water penetration
  • Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel and oil resistant outsole
  • Mono density sole unit
Material upper: Microfibre
Material sole: PU
These shoes do not utilise animal product or biproducts.
Certificaten: CE certified, EN ISO 20347 O2