Safety Boots, Slip-On, High, Classic Safety Wellington, S4
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FW94BKR48
PRTWFW94BKR48EA 29 EUR
PRTWFW94BKR48
Safety Boots, Slip-On, High, Classic Safety Wellington, S4
Schoenen en laarzen
Safety wellington is manufactured from virgin PVC/Nitrile in a seamless watertight construction. Ideal for use in a wide range of environments and industries.
  • Reinforced ankle protection
  • Chemical-resistant
  • Protective steel toecap
  • Anti-static footwear
  • Energy absorbing seat region
  • 100% waterproof to keep feet dry
  • SRC - Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel- and oil-resistant outsole
Classic safety wellington features include a 200 joule steel toecap, washable nylon lining, and kick off spurs.