Knee pad, KP40
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
KP40BKR
PRTWKP40BKREA 26 EUR
PRTWKP40BKR
Knee pad, KP40
Ergonomische steunen en hulpmiddelen Kniebeschermers
This strap-on gel knee pad offers tough protection for gruelling deep knee work. The thick gel adapts to the wearers knee, providing unrivalled comfort and protection.
  • Adjustable neoprene strap allows for greater flexibility
  • Abrasion resistant PVC shell ensures a longer useful lifespan
  • Specially designed shell gives great stability by distributing the weight evenly over a wide area
Certificaten: EN 14404:2004+A1:2010 Type 1 Level 1.