Standard ground joints
Leverancier: QUARTZTECHNISCHE WERKSTATTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1044 1047 1030 1045 1031 1033 1032 1046 1043
QUTW1044EA 80 EUR
QUTW1044 QUTW1047 QUTW1030 QUTW1045 QUTW1031 QUTW1033 QUTW1032 QUTW1046 QUTW1043
Standard ground joints
Adapters Adapters met NS-verbindingen
Connections, quartz glass, with standard ground joint according to DIN 12 249.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving high resistance to temperature changes.