Nitrogen flasks, round bottom with NS stopcock
Leverancier: QVF LABORTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
GSG01024H07 GSG01024H08 GSG01020H02 GSG01020H03 GSG01020H06 GSG01020H07
QVFLGSG01024H07EA 0 EUR
QVFLGSG01024H07 QVFLGSG01024H08 QVFLGSG01020H02 QVFLGSG01020H03 QVFLGSG01020H06 QVFLGSG01020H07
Nitrogen flasks, round bottom with NS stopcock
Kolven Stikstof kolven
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With conical ground joint, NS stopcock and inscription label.
Order Now
Specificaties
Environmentally Preferable