Ammoniumacetaat solution 5M

Leverancier: G Bioscienes
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Acetic acid ammonium salt

R012
R012EA 122 EUR
R012
Ammoniumacetaat solution 5M
Ammoniumacetaat
Formule: H₃CCOONH₄
MW: 77.08 g/mol
Smeltpunt: 114
Vlampunt: 136
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013066
CAS nummer: 631-61-8
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a80b41aae3d45f9440d931003e5299b07279d3a&vid=3644375d8ee47ec22264f175bd0d51c46214843e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR