Reichstein's substance S 98

Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 17alpha,21-Dihydroxy-4-pregnene-3,20-dione, Reichstein's Substance S, 11-Deoxy-17-hydroxycorticosterone

MQ200

R0500-1G
R0500-1GEA 66.5 EUR
R0500-1G
Reichstein's substance S 98
Cortexolone
Formule: C₂₁H₃₀O₄
MW: 346.47 g/mol
Smeltpunt: 208 °C (ontbinding, lit.)
Bewaar temperatuur: Room Temperature
MDL Nummer: MFCD00003662
CAS nummer: 152-58-9

Order Now

Specification Test Results

Appearance (Colour) White to Off-White
Appearance (Form) Powder
Infrared Spectrum Conforms to Structure
Purity (HPLC) ≥98 %
Solubility (Colour) Colorless to Faint Yellow
Solubility (Turbidity)
50 mg/mL, EtOH
Clear
Recommended Retest Period 6 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR