Fungal RNA kit, E.Z.N.A.®

Leverancier: OMEGA BIO-TEK
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

R6840-00 R6840-01
R6840-00EA 46.3 EUR
R6840-00 R6840-01
Fungal RNA kit, E.Z.N.A.®
Reagentia voor nucleïnezuren Nucleic Acid Purification Kits and Reagents

Modified from the E.Z.N.A.® Plant RNA kit, the E.Z.N.A.® Fungal RNA kit uses an improved buffer system that is more efficient for fungal samples. This kit provides convenient and rapid methods for the isolation of total RNA from a variety of fungal species. Rather, the method involves a simple and rapid precipitation step for removal of much of the polysaccharides and phenolic compounds commonly found in fungal tissues. In combination with HiBind® RNA spin columns, this method permits purification of high quality RNA from as much as 200 mg tissue. The system is also efficient enough to purify total RNA from as little as 10 mg of tissue or 100 cells. Purified RNA can be used for many downstream applications such as RT-PCR, northern blotting, Poly(A)+ mRNA purification, and differential display.


  • High quality RNA in less than 60 minutes
  • Optimised buffer system ensures reproducible results
  • No organic extractions
  • Purified DNA suitable for most downstream applications

Order Now

Specificaties

Omschrijving E.Z.N.A.® Fungal RNA Kit
Environmentally Preferable
Applicatie
Target molecule RNA
Refine target molecule Total RNA
Sample type Yeast/Fungi
Method / Format Column (silica-based)
Prep size Mini
Monstergrootte 100 mg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR