Test tubes
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3514030 3514040 3714010 3514031 3014030
RATI3514030EA 30.3 EUR
RATI3514030 RATI3514040 RATI3714010 RATI3514031 RATI3014030
Test tubes
Buisjes Reageerbuisjes
PS, clear or PP, translucent.
  • PS: Good thermal resistance. PP: Very good thermal resistance, autoclavable
  • PS: Good chemical resistance. PP: Very good chemical resistance
  • Robust, economical
Universal tubes for a wide range of applications.