Polyethylene glycol 8,000

Leverancier: G Bioscienes
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

RC-078 RC-077
RC-078EA 254 EUR
RC-078 RC-077
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Formule: H(OCH₂CH₂)nOH
Kookpunt: 250 °C (1013 hPa)
Dichtheid: 1,125 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01779615
CAS nummer: 25322-68-3
EINECS: 500-038-2
Merck Index: 12,07729

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR