Turbidity standards, Very low range 0 to 1 NTU
Leverancier: REAGECON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CRS-0.1-100 CRS-0.1-500 CRS-0.2-100 CRS-0.2-500 CRS-0.5-500 CRS-1-100 CRS-1-500 CRSR-0.1-100 CRSR-0.1-500 CRSR-0.2-100 CRSR-0.2-500 CRSR-0.4-100 CRSR-0.4-500 CRSR-0.5-100 CRSR-0.5-500 CRSR-1-100 CRSR-1-500 CRS-0.0-100 CRS-0.0-500
REGECRS-0.1-100EA 143 EUR
REGECRS-0.1-100 REGECRS-0.1-500 REGECRS-0.2-100 REGECRS-0.2-500 REGECRS-0.5-500 REGECRS-1-100 REGECRS-1-500 REGECRSR-0.1-100 REGECRSR-0.1-500 701-0048 REGECRSR-0.2-500 REGECRSR-0.4-100 REGECRSR-0.4-500 REGECRSR-0.5-100 REGECRSR-0.5-500 REGECRSR-1-100 REGECRSR-1-500 REGECRS-0.0-100 REGECRS-0.0-500
Turbidity standards, Very low range 0 to 1 NTU
Standaarden Standaarden voor troebelheidsbepaling
Turbidity standards for ratio and non-ratio instruments composed of suspended polymer microspheres.
  • Non toxic and non carcinogenic microspheres
  • Highly accurate with well defined spherical shape
  • Traceable to NIST
  • US EPA approved
  • Ready to use- particles stay in suspension