Conductivity standards, KCl, referenced at 25 °C

Leverancier: REAGECON
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CSKC10 CSKC100 CSKC1000 CSKC100M CSKC10M CSKC12880 CSKC150M CSKC20 CSKC200 CSKC200M CSKC20M CSKC300M CSKC3M CSKC5 CSKC50 CSKC500 CSKC50M CSKC5M CSKC84 CSKCL CSKCS CSKC136 CSKC350M CSKC500M CSKC13 CSKC2M CSKC80M CSKC450M CSKC238 CSKC300 CSKC400 CSKC111800 CSKC2500 CSKC25 CSKC185 CSKC250 CSKC30M CSKC400M CSKC60M CSKC40M CSKC1025
REGECSKC10EA 282 EUR
REGECSKC10 REGECSKC100 REGECSKC1000 REGECSKC100M REGECSKC10M REGECSKC12880 REGECSKC150M REGECSKC20 REGECSKC200 REGECSKC200M REGECSKC20M REGECSKC300M REGECSKC3M REGECSKC5 REGECSKC50 REGECSKC500 REGECSKC50M REGECSKC5M REGECSKC84 REGECSKCL REGECSKCS REGECSKC136 REGECSKC350M REGECSKC500M REGECSKC13 REGECSKC2M REGECSKC80M REGECSKC450M REGECSKC238 REGECSKC300 REGECSKC400 REGECSKC111800 REGECSKC2500 REGECSKC25 REGECSKC185 REGECSKC250 REGECSKC30M REGECSKC400M REGECSKC60M REGECSKC40M REGECSKC1025
Conductivity standards, KCl, referenced at 25 °C
Standaarden Geleidbaarheidsstandaarden

These conductivity standards are aqueous and so avoid the problems of matrix errors and high temperature dependency of nonaqueous conductivity standards.


  • With an accuracy of ±1,0% (except 1,3 µS)
  • Temperature dependence data, lot no. and expiry date are printed on product labels


These solutions standardised at 25 °C are directly traceable to N.I.S.T standard reference materials and are tested using an ISO 17025 accredited method.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR