EDTA disodium salt 0.01 mol/l (0.02 N) aqueous solution

Leverancier: REAGECON
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

ED20011 ED20015
REGEED20011EA 33.3 EUR
REGEED20011 REGEED20015
EDTA disodium salt 0.01 mol/l (0.02 N) aqueous solution
EDTA disodium salt
Formule: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00070672
CAS nummer: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR