Total organic carbon (TOC) standards
Leverancier: REAGECON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
RTOCK08 RTOCK09 RTOCK10 RTOCK11 RTOCK12 RTOCK13 RTOCK14 RTOCK15 RTOCK16 RTOCK17 RTOCK18 RTOCK19 RTOCK20 RTOCS01 RTOCS02 RTOCS03 RTOCS04 RTOCS05 RTOCS06 RTOCS07 RTOCS08 RTOCS09 RTOCS10 RTOCS11
REGERTOCK08EA 123 EUR
REGERTOCK08 REGERTOCK09 REGERTOCK10 REGERTOCK11 REGERTOCK12 REGERTOCK13 REGERTOCK14 REGERTOCK15 REGERTOCK16 REGERTOCK17 REGERTOCK18 REGERTOCK19 REGERTOCK20 REGERTOCS01 REGERTOCS02 REGERTOCS03 REGERTOCS04 REGERTOCS05 REGERTOCS06 REGERTOCS07 REGERTOCS08 REGERTOCS09 REGERTOCS10 REGERTOCS11
Total organic carbon (TOC) standards
Standaarden Standaarden voor milieuanalyse
  • Ready to use
  • Extended shelf life
Certificaten: In accordance with USP 643 and 1051 guidelines.