GC columns, ShinCarbon ST (packed and micropacked)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
80486-800 80486-810 80486-820 80486-840 19808 19809 19810 19043
REST80486-800EA 906 EUR
REST80486-800 REST80486-810 REST80486-820 REST80486-840 REST19808 REST19809 REST19810 REST19043
GC columns, ShinCarbon ST (packed and micropacked)
Kolommen voor chromatografie
These ShinCarbon ST columns are made of SilcoSmooth stainless steel material. Analyses oxygen, nitrogen, methane, carbon monoxide and carbon dioxide with one column at room temperature and separate permanent gases, including carbon monoxide and carbon dioxide, without cryogenic cooling.
  • Rapid separations of permanent gas/light hydrocarbon mixtures
  • Compatibile with most GC detectors - minimal bleed, minimal baseline rise
  • Preconditioned, less than 30 minutes to stabilise
  • Maximum temperature of 280/300 °C
ShinCarbon ST material, a high surface areaMore Product Information