Puller/inserter tool, CarboFrit®
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
21642
REST21642EA 30.5 EUR
REST21642
Puller/inserter tool, CarboFrit®
Uittrek- en invoeggereedschap
This puller/inserter tool has hook end for removing CarboFrit® inserts and a bent end (90°) for inserting CarboFrit® inserts.
  • Slender design
  • Easy to maneuver