Inlet liners, Sky®
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
23326 23305.1 23309.1 23434.5 23305.5 23312.1 23438.5 23336.5 23449.5 23312
REST23326EA 318 EUR
REST23326 REST23305.1 REST23309.1 REST23434.5 REST23305.5 REST23312.1 REST23438.5 REST23336.5 REST23449.5 REST23312
Inlet liners, Sky®
Benodigdheden voor chromatografie Injectiepoorten voor chromatografiesystemen
Sky® inlet liners with Sky® technology deactivation improve trace-level analysis that are suitable for Agilent GCs, Perkinelmer GCs, Shimadzu GCs and Thermo scientific GCs equipped with split/splitless inlets.
  • Increase accuracy and precision by preventing loss of sensitive analytes-even when using wool
  • Lower detection limits for a wide range of active compounds is achieved
  • Liner-to-liner reproducibility through consistent manufacturing and extensive testing