Vacuum gauges
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
24269 24270
REST24269EA 215 EUR
REST24269 REST24270
Vacuum gauges
Pompen Vacuümmeters
These vacuum gauges are made of stainless steel wetted surfaces with rear mount.
Suitable for passive air sampling kits.