PEEK tubing
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25065 25068 27751 25320
REST25065EA 0 EUR
REST25065 REST25068 REST27751 REST25320
PEEK tubing
Slangen Chromatography Tubing
Made of PEEK (Polyetheretherketone).
  • Less oxygen permeable and more temperature resistant than PTFE or Tefzel® tubing
  • Use with PEEK finger tight or flangeless fittings
This inert PEEK tubing replaces stainless steel, titanium, PTFE or Tefzel® tubing.