Tubing cutter, Clean-Cut
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25069
REST25069EA 69 EUR
REST25069
Tubing cutter, Clean-Cut
Snijders
This tubing cutter provides burr-free, perpendicular cuts that will not distort the tubing O.D. or close the I.D.
Use for PEEK, PTFE, Tefzel® and other polymeric tubing.