Sample delivery system, Resprep®
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
26250
REST26250EA 520 EUR
REST26250
Sample delivery system, Resprep®
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
This sample delivery system is used to transfer large volumes of low viscosity samples directly from a bottle to a solid phase extraction cartridge or to a disk on a vacuum manifold system for extraction or cleanup.
  • Compatible with 1, 3, 6, and 15 ml SPE cartridges and Diskcover-47 extraction disk holder
  • Tested to pH 1 to ensure no contaminants leach from system
  • Specified in EPA drinking water methods
Leveringsinformatie: Each unit consists of six PTFE transfer lines with a stainless steel weight on one end and a colour coded screw fitting with PETMore Product Information