Column heaters, Sidewinder™
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
26518 26516 26517
REST26518EA 0 EUR
REST26518 REST26516 REST26517
Column heaters, Sidewinder™
Verwarmers Kolomverwarmers
Column heaters and heater/cooler provides stable, accurate and reproducible results in HPLC analyses.
  • Light weight, compact design fits in small spaces
  • Utilise a 24 VDC input to energize the system
Certificaten: Meets the demanding electromagnetic emission standards of the new European Union Directives standards.