Test mix E1618 (13 components)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31613
REST31613EA 95 EUR
REST31613
Test mix E1618 (13 components)
Standaarden Standaarden voor de petroleumindustrie
Components in this mix (0,5 μl/ml or 0,05% volume/volume each) are at 10X the concentration of the final test solution specified in ASTM 1618 and ASTM 1387.
The mix consists of (C6) n-hexane, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C14) n-tetradecane, (C16) n-hexadecane, (C18) n-octadecane, (C20) n-eicosane, 2-ethyltoluene, 3-ethyltoluene, toluene, 1,2,4-trimethylbenzene, p-xylene.
Certificaten: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.