Calibration check mix, 8270 B/N (7 components)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31616
REST31616EA 51 EUR
REST31616
Calibration check mix, 8270 B/N (7 components)
Standaarden Standaarden voor milieuanalyse
Contains: Acenaphthene, benzo(a)pyrene, 1,4-dichlorobenzene, di-n-octyl phthalate, diphenylamine, fluoranthene, hexachlorobutadiene.
Certificaten: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.