Columns, MXT®-5
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
70205 70208 70220 70221 70222 70223 70224 70225 70226 70227 70228 70235 70236 70237 70238 70239 70240 70241 70250 70251 70252 70253 70254 70255 70257 70258 70267 70270 70277 70279 70281 70282 70283 70284 70285 70288 71821 71822 71823 71824 71825 70252-273 70255-273
REST70205EA 541 EUR
REST70205 REST70208 REST70220 REST70221 REST70222 REST70223 REST70224 REST70225 REST70226 REST70227 REST70228 REST70235 REST70236 REST70237 REST70238 REST70239 REST70240 REST70241 REST70250 REST70251 REST70252 REST70253 REST70254 REST70255 REST70257 REST70258 REST70267 REST70270 REST70277 REST70279 REST70281 REST70282 REST70283 REST70284 REST70285 REST70288 REST71821 REST71822 REST71823 REST71824 REST71825 REST70252-273 REST70255-273
Columns, MXT®-5
Kolommen voor chromatografie
These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.
  • Equivalent to USP G27, G36 phases
  • 4,5 inch standard coil diameter
General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.