Preparative HPLC columns, ultra PFP propyl
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9179557 9179517 9179518 9179558 9179559 9179567 9179568 9179578
REST9179557EA 2250 EUR
REST9179557 REST9179517 REST9179518 REST9179558 REST9179559 REST9179567 REST9179568 REST9179578
Preparative HPLC columns, ultra PFP propyl
Kolommen voor chromatografie
Ultra PFP (pentafluorophenyl) Propyl preparative columns are ideal for the retention and selectivity of charged bases, electronegative compounds, and amine-containing compounds.
  • More responsive to LC-MS as it is reliable and efficient with acidic mobile phases
  • Suitable for highly aqueous mobile phases and HILIC separations