HPLC columns, Viva PFP Propyl (USP L43)
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9519511 9519512 9519513 9519515 9519521 9519522 9519523 9519525 9519531 9519532 9519533 9519535 9519551 9519552 9519553 9519555 9519561 9519562 9519563 9519565 9519571 9519572 9519573 9519575 9519512-700 9519513-700 9519515-700 951951E 9519522-700 9519523-700 9519525-700 951952E 9519532-700 9519533-700 9519535-700 951953E 9519552-700 9519553-700 9519555-700 951955E 9519562-700 9519563-700 951950210 951950212 9519565-700 951956E 9519572-700 9519573-700 9519575-700 951957E
REST9519511EA 678 EUR
REST9519511 REST9519512 REST9519513 REST9519515 REST9519521 REST9519522 REST9519523 REST9519525 REST9519531 REST9519532 REST9519533 REST9519535 REST9519551 REST9519552 REST9519553 REST9519555 REST9519561 REST9519562 REST9519563 REST9519565 REST9519571 REST9519572 REST9519573 REST9519575 REST9519512-700 REST9519513-700 REST9519515-700 REST951951E REST9519522-700 REST9519523-700 REST9519525-700 REST951952E REST9519532-700 REST9519533-700 REST9519535-700 REST951953E REST9519552-700 REST9519553-700 REST9519555-700 REST951955E REST9519562-700 REST9519563-700 REST951950210 REST951950212 REST9519565-700 REST951956E REST9519572-700 REST9519573-700 REST9519575-700 REST951957E
HPLC columns, Viva PFP Propyl (USP L43)
Kolommen voor chromatografie
Viva PFP Propyl columns with a pentafluorophenyl propyl phase are ideal for retention and selectivity of charged bases, electronegative compounds and amine-containing compounds.
  • Rugged, spherical particles with 300 Å pore size
  • Excellent for separating peptides or proteins
  • High proportion of pore/surface area available to large molecules
These are reliable and efficient with acidic mobile phases. The versatile column is suitable for highly aqueous mobile phases and HILIC separations.