Fittings, RheFlex®, Rheodyne®
Leverancier: IDEX RHEODYNE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6000-051 6000-054 6000-055 6000-066 6000-066GN 6000-066RD 6000-066WH 6000-078 6000-090 6000-116 6000-210 6000-211 6000-251 6000-254 6000-254GN 6000-254RD 6000-254YL 6000-263 6000-264 6000-265 6000-278 6000-280RD 6000-282 6000-282BL 6000-282GN 6000-282MC 6000-282RD 6000-282YL
554-2029EA 67.9 EUR
554-2029 554-2030 554-2031 554-2032 RHEO6000-066GN RHEO6000-066RD RHEO6000-066WH RHEO6000-078 554-2033 RHEO6000-116 RHEO6000-210 RHEO6000-211 RHEO6000-251 RHEO6000-254 RHEO6000-254GN RHEO6000-254RD RHEO6000-254YL RHEO6000-263 RHEO6000-264 RHEO6000-265 RHEO6000-278 RHEO6000-280RD RHEO6000-282 RHEO6000-282BL RHEO6000-282GN RHEO6000-282MC RHEO6000-282RD RHEO6000-282YL
Fittings, RheFlex®, Rheodyne®
Fittingen en connectoren
RheFlex® tube fittings provide inert, non-metal connections, and can be used with all Rheodyne injectors, valves and sample loops.
The fitting holds with finger tightening on a variety of plastic and metal tubing.
Order Now
Specificaties
Environmentally Preferable