Screw Cap, PP
Leverancier: RIXIUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8-3000-28-W
RIXI8-3000-28-WEA 0.3 EUR
RIXI8-3000-28-W
Screw Cap, PP
Sluitingen Flesdoppen
Closure made of PE white 0/001 with inner cone for thread 28 for round bottle 250 to 1000 ml.