Filtrartion crucibles for dietry fibre extractions, VitraPOR®
Leverancier: ROBU GLASFILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20600 20601 20602 20603 20604
ROBU20600EA 217 EUR
ROBU20600 ROBU20601 ROBU20602 ROBU20603 ROBU20604
Filtrartion crucibles for dietry fibre extractions, VitraPOR®
Kroezen
These filltration crucibles are suitable for crude fibre extraction.
  • Fit many current systems
  • Numbered
The VitraPOR® filter discs of the CFE crucibles have an improved sintering hardness to withstand the chemical load in the analytical processes.