Anti-PDIA2 Rabbit Polyclonal Antibody

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

100-401-G29
ROCK100-401-G29EA 821 EUR
ROCK100-401-G29
Anti-PDIA2 Rabbit Polyclonal Antibody
Antilichamen
Anti-PDI Antibody is suitable for use in WB, IP, IF microscopy and IHC. Expect a band approximately ~58kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:4000;

Type: Primary
Antigen: PDIA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog, Hamster, Monkey, Guinea Pig, Bovine, Sheep, Pig, Xenopus, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR