Anti-LPO Sheep Polyclonal Antibody

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

100-601-146
ROCK100-601-146EA 2060 EUR
ROCK100-601-146
Anti-LPO Sheep Polyclonal Antibody
Antilichamen
Primary Sheep Anti-LACTOPEROXIDASE (Bovine Milk) Reacts with Pan

Recommended Dilutions: ELISA: 1:18000 - 1:75000; Western Blot: 1:2000 - 1:10000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: LPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Bovine, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR