TrueBlot® IP/Western Blot
Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
13-8817-82 00-8811-25 13-8817-80 88-1488-31 88-1688-31 88-7788-31
ROCK13-8817-82EA 420 EUR
ROCK13-8817-82 ROCK00-8811-25 ROCK13-8817-80 ROCK88-1488-31 ROCK88-1688-31 ROCK88-7788-31
TrueBlot® IP/Western Blot
Blotkits
IP Western blots provide highly specific results, yet often suffer from heavy/light chain blotting, contamination, and ongoing interference.
TrueBlot® reagents enable you to generate clear, best-quality data in your Immunoprecipitation and Western Blot protocols. Available in several options, from IP Beads alone, to complete IP/Western Blot kits from goat, mouse, rabbit or sheep.
TrueBlot® products solve nearly all of these problems through increased sensitivity, less background noise, and enhanced accuracy.