Anti-HCN2 Mouse Monoclonal Antibody (Unconjugated) [clone: S71-37]

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

200-301-F41
ROCK200-301-F41EA 841 EUR
ROCK200-301-F41
Anti-HCN2 Mouse Monoclonal Antibody (Unconjugated) [clone: S71-37]
Antilichamen
Anti-HCN2 Antibody is suitable for use in WB, IP and IHC. Expect a band approximately ~95kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: HCN2
Clonality: Monoclonal
Clone: S71-37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR