Anti-RHO Mouse Monoclonal Antibody (Unconjugated) [clone: 4D2]

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

200-301-G36
ROCK200-301-G36EA 841 EUR
ROCK200-301-G36
Anti-RHO Mouse Monoclonal Antibody (Unconjugated) [clone: 4D2]
Antilichamen
Anti-Rho Antibody is recommended for use in IF microscopy, IHC, IP, WB, and ELISA. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:1000;

Type: Primary
Antigen: RHO
Clonality: Monoclonal
Clone: 4D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mammalian, Vertebrates, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR