Anti-TRPM7 Mouse Monoclonal Antibody (Unconjugated) [clone: S74-25]

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

200-301-G57
ROCK200-301-G57EA 841 EUR
ROCK200-301-G57
Anti-TRPM7 Mouse Monoclonal Antibody (Unconjugated) [clone: S74-25]
Antilichamen
Anti-TRPM7 Antibody is suitable for use in WB, IP, and Immunofluorescence microscopy. Antibody is provided in PBS pH 7.4. Expect a band approximately ~220kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: TRPM7
Clonality: Monoclonal
Clone: S74-25
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR