Anti-CD80 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 2D10]

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

200-302-N74
ROCK200-302-N74EA 699 EUR
ROCK200-302-N74
Anti-CD80 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 2D10]
Antilichamen
Anti-CD80 is useful for Flow Cytometry, immunochemistry, and Western Blot. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells;
Do not freeze!

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 2D10
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR