Anti-AKT1 Mouse Monoclonal Antibody (Biotin) [clone: 14E5.A2.B2.H9]

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

200-306-I51
ROCK200-306-I51EA 0 EUR
ROCK200-306-I51
Anti-AKT1 Mouse Monoclonal Antibody (Biotin) [clone: 14E5.A2.B2.H9]
Antilichamen
Anti-AKT3 BIOTIN Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT3 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 14E5.A2.B2.H9
Conjugation: Biotin
Epitope: Center
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse, Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR