Anti-GST Mouse Monoclonal Antibody (DyLight® 488) [clone: 3D4]

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

200-341-200
ROCK200-341-200EA 791 EUR
ROCK200-341-200
Anti-GST Mouse Monoclonal Antibody (DyLight® 488) [clone: 3D4]
Antilichamen
Type: Primary
Antigen: GST
Clonality: Monoclonal
Clone: 3D4
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype:
Reactivity: pan
Order Now

Specificaties

antigeen symbool GST
Type antilichaam Primair,Glutathione-S-Transferase
Environmentally Preferable
Type antilichaam Primair,Glutathione-S-Transferase
Clonality Monoklonaal
Conjugatie DyLight® 488
Clone 3D4
Reactivity Multiple
Host Mouse
Isotype IgG1 kappa
Western blot Yes
ELISA Yes
ImmunoChemie Yes
ImmunoFluorescentie Yes
Gelverschuiving Yes
ChIP Yes
Neutralisation Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR