Anti-TMBIM1 Rabbit Polyclonal Antibody

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

200-401-A57
ROCK200-401-A57EA 821 EUR
ROCK200-401-A57
Anti-TMBIM1 Rabbit Polyclonal Antibody
Antilichamen
This Protein A purified antibody has been tested for use in ELISA, immunochemistry, and western blotting.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 35 kDa in size corresponding to TMBIM-1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500 - 1:2000; Immunochemistry: 10µg/ml;

Type: Primary
Antigen: TMBIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Specificaties

antigeen symbool TMBIM1
Environmentally Preferable
Type antilichaam Primair,transmembrane BAX inhibitor motif containing 1
Antigeen synoniemen MSTP100|RECS1|LFG3|PP1201|MST100
Type antilichaam Primair,transmembrane BAX inhibitor motif containing 1
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Gen-ID 64114
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Immunogeen This Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant Protein corresponding to the amino terminus of human TMBIM1 Protein.
Formaat Antibody fraction
Formulier Liquid (sterile filtered)
Purification Protein A purified
Kruisadsorptie No
Opslagbuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Opslagtemperatuur –20 °C
Shipping temperature Dry Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR