Anti-CD86 Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: GL-1]

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

200-526-N90
ROCK200-526-N90EA 797 EUR
ROCK200-526-N90
Anti-CD86 Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: GL-1]
Antilichamen
Anti-CD86 is useful for immunochemistry, Immunofluorescence, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: GL-1
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Specificaties

antigeen symbool CD86
Environmentally Preferable
Type antilichaam Primair,CD86 molecule
Antigeen synoniemen CLS1|B70|B7.2|Ly-58|Cd28l2|ETC-1|MB7|TS/A-2|B7-2|B7|MB7-2|Ly58
Type antilichaam Primair,CD86 molecule
Clonality Monoklonaal
Conjugatie APC (Allophycocyanin)
Clone GL-1
Reactivity Mouse
Host Rat
Gen-ID 12524
Isotype IgG2a kappa
ImmunoChemie Yes
ImmunoFluorescentie Yes
Flowcytometrie Yes
Immunogeen Anti-CD86 Antibody (Monoclonal) was produced by repeated immunizations with CD86 antigen.
Formaat Antibody fraction
Formulier Liquid (sterile filtered)
Purification affinity-purified
Concentratie 0,2 mg/ml by UV absorbance at 280 nm
Kruisadsorptie none
Opslagbuffer 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,1% Gelatin
Opslagtemperatuur Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR