Anti-IgG F(c) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

205-7203
ROCK205-7203EA 448 EUR
ROCK205-7203
Anti-IgG F(c) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: F(c)
Host: Donkey
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR