Anti-BIN1 Mouse Monoclonal Antibody (Unconjugated) [clone: 12A]

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

209-301-E62S
ROCK209-301-E62SEA 226 EUR
ROCK209-301-E62S
Anti-BIN1 Mouse Monoclonal Antibody (Unconjugated) [clone: 12A]
Antilichamen
Type: Primary
Antigen: BIN1
Clonality: Monoclonal
Clone: 12A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR